Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta Actin Monoclonal Antibody (2D1D10), THE™, GenScript™

Catalog No. p-7222679
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
200 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-272-953 100 μg
50-272-955 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-272-953 Supplier Genscript Corporation Supplier No. A00702100
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute the lyophilized antibody with deionized water to the final concentration of 0.5 mg/mL. beta-actin is one of the six different actin isoforms that have been identified. The actin molecules found in cells of various species and tissues tend to be very similar in their immunological and physical properties. As a consequence, it has been difficult to produce potent antisera against this protein. Therefore, the availability of monoclonal antibodies to beta-actin provides a useful and specific tool in the study of the intracellular distribution of β-actin and the static and dynamic aspects of the cytoskeleton.

beta Actin Monoclonal antibody specifically detects beta Actin in Bovine, Chicken, Fish, Goat, Hamster, Human, Mouse, Porcine, Rabbit samples. It is validated for ELISA, Western Blot

Specifications

Antigen beta Actin
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2D1D10
Conjugate Unconjugated
Formulation PBS with 1% BSA and 0.02% sodium azide; pH 7.4
Gene ACTB
Gene Accession No. P29751, P48975, P60706, P60709, P60710, P60712, Q6QAQ1
Gene Alias 0610041G09Rik; AA959943; AAT6; ACT; Act4; Act-4; ACT-5; ACTA; acta1; acta1b; ACTA2; Acta-2; ACTA3; actb; actb.L; actb1; actba; ACTC; actc1; Actc-1; actc1b; ACTE; Actg; ACTG1; Actg2; ACTGE; actin; actin alpha 1; actin alpha 1, skeletal muscle; actin alpha 1, skeletal muscle b; actin alpha 2, smooth muscle; actin alpha cardiac; actin alpha cardiac 1; actin alpha cardiac muscle 1; actin alpha cardiac muscle 1b; actin beta; actin gamma 1; actin gamma 2, smooth muscle; actin, alpha 1, skeletal muscle; actin, alpha 1b, skeletal muscle; actin, alpha 2, smooth muscle, aorta; Actin, alpha cardiac muscle 1; Actin, alpha cardiac muscle 1, intermediate form; actin, alpha cardiac muscle 1b; actin, alpha skeletal muscle; Actin, alpha skeletal muscle, intermediate form; actin, alpha, cardiac 1; actin, alpha, cardiac muscle; actin, alpha, cardiac muscle 1; actin, alpha, vascular smooth muscle; actin, aortic smooth muscle; Actin, aortic smooth muscle, intermediate form; actin, beta; actin, beta 1; actin, beta L homeolog; actin, beta, cytoplasmic; actin, cytoplasmic 1; Actin, cytoplasmic 1, N-terminally processed; Actin, cytoplasmic 2; Actin, cytoplasmic 2, N-terminally processed; actin, gamma 1; actin, gamma 2, smooth muscle, enteric; actin, gamma, cytoplasmic 1; actin, gamma-enteric smooth muscle; Actin, gamma-enteric smooth muscle, intermediate form; actin-like protein; Actl; ACTL3; Acts; ACTSA; ACTSG; Actsk-1; Actvs; Actx; AL023024; alpha actin 1; alpha-actin cardiac; alpha-actin-1; Alpha-actin-2; alpha-actin-3; alphac-actin; Alpha-cardiac actin; alphaSMA; alpha-smooth muscle actin; ASD5; ASMA; a-SMA; Atcb; Bact; Bact; actin; B-actin; bactin1; bactin1 protein; bactzf; B-ACTZF; beta actin; beta cytoskeletal actin; beta-actin; beta-actin FE-3; beta-actin-1; BRWS1; BRWS2; Cap Z; capZ beta; cardiac muscle alpha actin 1; cardiofunk; cell growth-inhibiting gene 46 protein; Cfk; CFTD; CFTD1; CFTDM; CMD1R; CMH11; cytoplasmic 1; cytoplasmic beta-actin; cytoskeletal beta actin; cytoskeletal gamma-actin; cytoskeletal protein; cytoskeleton actin; deafness, autosomal dominant 20; deafness, autosomal dominant 26; DFNA20; DFNA26; E430023M04Rik; E51; epididymis luminal protein 176; fa27h01; fb83f06; gamma actin; gamma non-muscle actin; gamma-2-actin; Gamma-actin; gamma-enteric smooth muscle actin; GIG46; HEL-176; hm:zeh0631; I79_002310; I79_013242; I79_019066; LVNC4; MPFD; MYMY5; NEM1; NEM2; NEM3; nemaline myopathy type 3; PS1TP5-binding protein 1; PS1TP5BP1; SHPM; similar to beta actin; skeletal alpha actin; skeletal alpha1 actin; sma; SMalphaA; SMGA; smooth muscle alpha-actin; smooth muscle gamma-actin; vascular smooth muscle alpha-actin; VSCM; wu:fa27h01; wu:fb63d03; wu:fb83f06; wu:fd18f05; XELAEV_18045052mg; zeh0631
Host Species Mouse
Immunogen A synthetic peptide DDDIAALVVDNGSG coupled to KLH
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100009272, 100689477, 101844587, 102179831, 11461, 280979, 396526, 414396, 60
Target Species Bovine, Chicken, Fish, Goat, Hamster, Human, Mouse, Pig, Rabbit
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Lyophilized
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.