Learn More
LI COR BIOTECH LLC Actin Rabbit Monoclonal Antibody for Normalization (100 L)
Supplier:  LI COR BIOTECH LLC 92642210
Encompass_Preferred
The β-actin primary antibody can be used as an internal loading control (ILC) for normalization. The expression of β-actin, or any housekeeping protein (HKP), should be validated to ensure that its expression does not change under experimental conditions. Once validated, β-actin primary antibodies can be used for the detection of β-actin when performing two-color detection. Detect β-Actin Rabbit Primary Antibody with IRDye Goat anti-Rabbit or IRDye Donkey anti-Rabbit secondary antibodies.
Catalog No. NC9872617
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.