Learn More
REVMAB BIOSCIENCES USA INC β-Actin [RM112], Rabbit Monoclonal Antibody
Supplier:  REVMAB BIOSCIENCES USA INC 31101300
Encompass_Preferred
Anti-β-Actin rabbit monoclonal antibody, Clone RM112 (100 uL) ; WB, IP, ICC, IHC, FC, ELISA validated; Product info: www.revmab.com
Catalog No. NC1729073
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.