Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beta Actin Recombinant Rabbit Monoclonal Antibody (4H1), Proteintech
SDP

Catalog No. 502334330 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-233-4330 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-233-4330 Supplier Proteintech Group Inc Supplier No. 811151RR100UL
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Beta ActinRecombinant Monoclonal antibody specifically detects Beta Actin in Human, Mouse, Pig, Rat samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen Beta Actin
Applications Western Blot
Classification Recombinant Monoclonal
Clone 4H1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Accession No. P60709
Gene Alias ACTB; Actx; beta-actin; BRWS1; E430023M04Rik; PS1TP5BP1
Host Species Rabbit
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60
Target Species Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.