Learn More
AnaSpec Beta - Amyloid (1 - 40), HiLyteᵀᴹ Fluor 647 - labeled, Human, 0.1mg
Supplier:  AnaSpec AS60493
Encompass
Sequence: HiLyte™ Fluor 647-DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVV, Molecular Weight 5315.4, Beta - Amyloid (1 - 40), HiLyteᵀᴹ Fluor 647 - labeled, Human, 0.1mg
Catalog No. NC0904817