Learn More
AnaSpec Beta - Amyloid (1 - 42), HiLyteᵀᴹ Fluor 647 - labeled, Human, 0.1mg
Supplier:  AnaSpec AS64161
Encompass
Beta-Amyloid (1-42), HiLyte(TM) Fluor 647-labeled, Human[HiLyte(TM) Fluor 647-DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIA] ; MW 5449.4; (0.1 mg) This beta-amyloid (1-42) peptide is labeled on the N-terminus with HiLyte(TM) Fluor 647, Abs/Em = 649/674 nm.
Catalog No. NC0904818