Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta Amylase Polyclonal Antibody, HRP, Rockland™
SDP

Catalog No. p-7190652
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1096 100 μg
50-199-1097 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-1096 Supplier Rockland Immunochemicals Supplier No. 20043470100
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Beta Amylase is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Peroxidase, anti-Rabbit Serum as well as purified and partially purified Beta Amylase [Sweet Potato]. Cross reactivity against Beta Amylase from other tissues and species may occur but have not been specifically determined.

Beta Amylase is an enzyme found in the seeds of higher plants and in sweet potatoes. It hydrolyses 1,4 alpha D glucosidic linkages in polysaccharides to remove successive maltose units from the non reducing ends of the chains. Used in the structural analysis of starch and glycogen.

Specifications

Antigen beta Amylase
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate HRP
Formulation 0.02M potassium phosphate with 0.15M NaCl, 10 mg/mL BSA and 0.01% gentamicin sulfate; pH 7.2
Gene Alias ß-amylase
Host Species Rabbit
Immunogen Beta Amylase [Sweet Potato]
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Plant
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.