Learn More
Apexbio Technology LLC Beta-Amyloid (1-11) 190436-05-6 10mg
SDP
Supplier:  Apexbio Technology LLC A100210
Encompass_Preferred
Beta-Amyloid (1-11) (CAS 190436-05-6) is a synthetic peptide fragment derived from the N-terminus of the amyloid precursor protein comprising the amino acid sequence H-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-OH This peptide represents a portion of the beta-amyloid (A ) sequence implicated in amyloidogenic pathways associated with neurodegenerative processes particularly Alzheimer s disease Beta-Amyloid (1-11) is utilized in research to investigate amyloid aggregation dynamics immunogenic epitope mapping within the A region and the biochemical properties of amyloidogenic peptides It serves as a molecular tool in studies examining the early stages of A processing clearance and interactions with cellular components
Catalog No. 50-000-95130
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.