Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ beta Amyloid (1-42) Polyclonal Antibody, Biotin
GREENER_CHOICE

Catalog No. 443449
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-344-9 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-344-9 Supplier Invitrogen™ Supplier No. 443449
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This beta Amyloid (1-42) antibody (44-3449) is purified from rabbit serum by epitope-specific affinity chromatography. Any reactivity towards both full length Abeta 40 and Abeta 43 protein has been eliminated through a series of preabsorption steps. This antibody shows no significant cross-reactivity against Abeta 40 or Abeta 43.

Beta amyloid is a 40-43 amino acid peptide cleaved from amyloid precursor protein by a protease, gamma-secretase. Beta Amyloid (1-40) (Amyloid beta 40 or Abeta 40) together with beta Amyloid (1-42) (Amyloid beta 42 or Abeta 42) are two major C-terminal variants of the amyloid beta protein, constituting the majority of amyloid beta proteins. Both the Amyloid beta 40 and Amyloid beta 42 alloforms are major components of the extracellular plaques found in Alzheimer's disease brain tissue.
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta Amyloid (1-42)
Applications Dot Blot, Functional Assay, Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate Biotin
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene APP
Gene Accession No. P05067
Gene Alias A beta 1 42; A ß 1 42; A4; AAA; AB42; Abeta; Abeta40; Abeta42; ABPP; AD1; Adap; AG; AICD-50; AICD-57; AICD-59; AID(50); AID(57); AID(59); Alpha-CTF; Alpha-secretase C-terminal fragment; Alzheimer disease amyloid A4 protein homolog; alzheimer disease amyloid protein; Amyloid; amyloid A4; Amyloid b; Amyloid beta; amyloid beta (A4) precursor protein; Amyloid beta 1 42; amyloid beta A4 protein; amyloid beta precursor protein; Amyloid intracellular domain 50; Amyloid intracellular domain 57; Amyloid intracellular domain 59; Amyloid precursor protein; Amyloid ß; Amyloid-beta A4 protein; Amyloid-beta precursor protein; Amyloid-beta protein 40; Amyloid-beta protein 42; Amyloidogenic glycoprotein; APP; APP-C57; APP-C59; APP-C99; APPI; appican; Aß 1 42; B APP42; BAPP42; beta-amyloid peptide; beta-amyloid peptide(1-40); beta-amyloid peptide(1-42); beta-amyloid precursor protein; betaApp; Beta-APP40; BetaAPP42; Beta-APP42; Beta-CTF; Beta-secretase C-terminal fragment; C31; C80; C83; C99; Cerebral vascular amyloid peptide; CTFgamma; Cvap; E030013M08Rik; Gamma-CTF(50); Gamma-CTF(57); Gamma-CTF(59); Gamma-secretase C-terminal fragment 50; Gamma-secretase C-terminal fragment 57; Gamma-secretase C-terminal fragment 59; N-APP; P3(40); P3(42); peptidase nexin-II; PN2; PN-II; PreA4; protease nexin II; protease nexin-II; S-APP-alpha; S-APP-beta; Soluble APP-alpha; Soluble APP-beta; testicular tissue protein Li 2; ß APP42; ßAPP42
Gene Symbols APP
Host Species Rabbit
Immunogen 1-42 of Amyloid Beta.
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 351
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.