Learn More
CPC Scientific H-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-Ile-Ala-OH 0.5MG
SDP
Supplier:  CPC Scientific AMYD003A
Encompass
42-residue peptide believed to be the major subunit of the vascular and plaque filaments in individuals with Alzheimer's disease, elderly people and patients with Trisomy 21 (Down's Syndrome). Inactive control.SEQUENCE: H-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-Ile-Ala-OHONE-LETTER SEQUENCE: DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIAMOLECULAR FORMULA: C203H311N55O60S1MOLECULAR WEIGHT: 4514.04STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [107761-42-2]SYNONYMS: Amyloid beta-Protein (1-42), Abeta42RESEARCH AREA: Alzheimer's DiseaseREFERENCES:J.Kang et al., Nature, 325, 733 (1987)D.Goldgaber et al., Science, 235, 877 (1987)J.Murrell et al., Science, 254, 97 (1991)Neurobiology of Aging, 13, No.5 (1992)G.Bitan et al., Proc. Natl. Acad. Sci. USA, 100, 330 (2003)A.Jan et al., Nat. Protoc., 5, 1186 (2010)W.B.Stine et al., Methods Mol. Biol. , 670
Catalog No. 50-210-8649
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.