Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ beta Amyloid 42 Peptide
SDP

Catalog No. p-7249215 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171974 100 μg
NB171976 500 μg
NB171975 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. NB171974 Supplier Novus Biologicals™ Supplier No. NBP318318100UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

A recombinant protein corresponding to human beta amyloid 42

Amino Acid Sequence: DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIA

Specifications

For Use With (Application) Electron Microscopy, Immunomicroscopy, In vitro Assay, In vivo Assay, Western Blot
Formulation Dry powder. SeeReconstitution Instructions for re-suspension instructions/protocol.
Gene ID (Entrez) 351
Molecular Weight (g/mol) M.W. Theoretical: 4.5 kDa
Quantity 100 μg
Storage Requirements Store at -70C. Avoid freeze-thaw cycles.
Gene Alias AAA, Abeta, ABPP, AD1, alpha-sAPP, Amyloid-beta precursor protein, APPI, Beta-Amyloid Peptide(1-42), CTFgamma, CVAP, PN2, PreA4
Gene Symbol APP
Purity or Quality Grade >85%
Protein beta Amyloid 42
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.