Learn More
Specifications
Specifications
| Gene ID (Entrez) | 351 |
| Species | Human |
| Purity | >98%, by HPLC |
| Content And Storage | Store at -70°C. Avoid freeze-thaw cycles. |
| Gene Alias | AAA, Abeta, ABPP, AD1, alpha-sAPP, Amyloid-beta precursor protein, APPI, Beta-Amyloid Peptide(1-42), CTFgamma, CVAP, PN2, PreA4 |
| Gene Symbol | APP |
| Molecular Weight (g/mol) | 4.5 kDa |
| Quantity | 5 x 100 ug Vials |
| Storage Buffer | Dry powder. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
