Learn More
AnaSpec Beta - Amyloid Peptide (1 - 42), mouse, rat, 0.5mg
Supplier:  AnaSpec AS25381
Encompass
Beta - Amyloid Peptide (1 - 42), mouse, rat, 0.5mg, Sequence: DAEFGHDSGFEVRHQKLVFFAEDVGSNKGAIIGLMVGGVVIA, Molecular Weight 4418,
Catalog No. NC0956788