Learn More
Abcam beta-Amyloid Peptide (31-35), 1MG
Supplier:  Abcam AB1209741MG
Encompass_Preferred
MW 545.74 g/mol, Purity >95%. Amyloidogenic peptide. Induces apoptosis/neurodegeneration in vitro. These effects are orchestrated by the oxidation status of methionine 35. Involves a number of signaling pathways including JNK, caspases and PKA.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7075