Learn More
Description
MA1-183 detects beta Arrestin 1 protein from human samples at approximately 55 kDa.
Specifications
Specifications
| Antigen | beta Arrestin 1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 4E11B1 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene | ARRB1 |
| Gene Accession No. | P49407 |
| Gene Alias | 1200006I17Rik; ARB1; ARR1; ARRB1; ARRB1 protein; arrdc1; arrestin; arrestin 2; arrestin beta 1; arrestin beta-1; arrestin, beta 1; arrestin-2; AW208571; BARRES; beta-arrestin; beta-arrestin1; beta-arrestin-1; G430100A01Rik; Non-visual arrestin-2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
