Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ beta-Arrestin 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA1732 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1732 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1732 Supplier Invitrogen™ Supplier No. PA1732
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PA1-732 detects beta-Arrestin 2 in rat, human and bovine samples. PA1-732 has been successfully used in Western blot procedures. By Western blot, this antibody detects ~49 kDa and ~47 kDa proteins representing pan Arrestin in rat and bovine retina as well as rat brain samples. The PA1-732 immunogen is a synthetic peptide corresponding to residues C D(384) D I V F E D F A R L R L K(397) of human beta-arrestin 2. This peptide (Cat. # PEP-281) is available for use in neutralization and control experiments.

Beta-arrestin-2, or ARRB2, is an adaptor protein belonging to the arrestin family with an N- arrestin domain and a C- arrestin domain. ARRB2 is a regulatory protein involved in heterotrimeric G protein-coupled receptor desensitization and is known to regulate beta2-adrenergic receptor (beta2AR) function by enhancing beta2AR mediated nuclear translocation of ERK. ARRB2 binds to phosphorylated beta2ARs, thereby causing a significant impairment of their capacity to activate G(S) proteins. Along with AIP4, ARRB2 acts as an endosomal sorting molecule that mediates CXCR4 entry into a degradative pathway. It may also be involved in hormone-specific desensitization of TSH receptors. ARRB2 is a Ca2+ binding protein of the retinal outer segments and binds to P-rhodopsin and is also involved in synaptic transmission in photoreceptor cells. It is widely expressed in most of the tissues, especially neuronal tissues and spleen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta-Arrestin 2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene ARRB2
Gene Accession No. P29067, P32120, P32121, Q91YI4
Gene Alias AI326910; ARB2; ARR2; ARRB2; arrestin 3; arrestin beta 2; arrestin beta-2; arrestin, beta 2; arrestin-3; AW122872; BARR2; BARRES; beta arr2; beta-arrestin 2 long isoform; beta-arrestin 2 short isoform; beta-arrestin-2; Non-visual arrestin-3
Gene Symbols ARRB2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues C D(384) D I V F E D F A R L R L K(397) of human beta-arrestin 2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216869, 25388, 281638, 409
Target Species Human, Mouse, Rat, Bovine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.