Learn More
POLYCARBIN INC 15 mL Centrifuge Tube CS500, Racked (Sterile)
SDP
Supplier:  POLYCARBIN INC PC010201G
Encompass_Preferred
Polycarbin's racked 15 mL conical tubes are made using 20% circular economy, post-consumer recycled content. Each lot of tubes is sterile. Certified DNase, RNase, and Pyrogen free. 50 tubes/rack, 10 racks/case, 500 tubes/case. Centrifugation Speed 12,000 x g. Temperature Range of -196°C to 100°C. Manufactured in a ISO 9001 compliant facility.
Catalog No. NC2213711
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.