Learn More
OriGene Beta-Catenin (CTNNB1) Protein, 100 µg, High Purity Derived from HEK293T Cells
SDP
Supplier:  OriGene TP314557M
Encompass_Preferred
Beta-Catenin (CTNNB1) Protein is a highly purified recombinant protein from OriGene Technologies, engineered for scientific research applications. It is derived from human HEK293T cells and optimized for in vitro assays, such as cell culture applications. With a quantity of 100 µg and designed for long-term stability at -80°C, this protein ensures high quality and specificity for reliable results in various research settings.
Catalog No. NC2198440
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.