Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ beta COP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA1068
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1068 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1068 Supplier Invitrogen™ Supplier No. PA1068
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-068 detects coatomer-protein I beta' (COPI beta) from rat cells. PA1-068 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~115 kDa protein representing COP beta from PC-12 cell extract. A non-specific band at ~100 kDa may be seen. PA1-068 immunogen is a synthetic peptide corresponding to residues K(494) V L A A Q E T H E G V T E D G I E(511) of rat beta-COP. The PA1-068 immunizing peptide (Cat. # PEP-126) is available for use in neutralization and control experiments.

This gene encodes a protein subunit of the coatomer complex associated with non-clathrin coated vesicles. The coatomer complex, also known as the coat protein complex 1, forms in the cytoplasm and is recruited to the Golgi by activated guanosine triphosphatases. Once at the Golgi membrane, the coatomer complex may assist in the movement of protein and lipid components back to the endoplasmic reticulum. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta COP
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1 mg/mL BSA and 0.05% sodium azide
Gene Copb1
Gene Accession No. P23514
Gene Alias 2610019B04Rik; beta coat protein; beta-coat protein; Beta-COP; coatomer protein complex subunit beta 1; coatomer protein complex, subunit beta 1; Coatomer subunit beta; COPB; COPB1; DKFZp761K102; FLJ10341; FLJ46444; FLJ57957; I79_013776; MSTP026; Rack2; receptor for activated C-kinase-2
Gene Symbols Copb1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues K(494) V L A A Q E T H E G V T E D G I E(511) of rat beta-COP.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114023
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.