Learn More
Creative Biomart Tilactase; Beta-D-galactosidase
Supplier:  Creative Biomart THP0026
Encompass_Preferred
The product is a beta-D-galactosidase obtained from Aspergillus oryzae. The beta-D-galactosidase us a large monomeric multi-domain enzyme of 985 residues that presents a catalytic (alpha/beta)8-barrel domain.
Catalog No. 50-208-4077
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.