Learn More
CPC Scientific H-Asp-His-Tyr-Asn-Cys-Val-Ser-Ser-Gly-Gly-Gln-Cys-Leu-Tyr-Ser-Ala-Cys-Pro-Ile-Phe-Thr-Lys-Ile-Gln-Gly-Thr-Cys-Tyr-Arg-Gly-Lys-Ala-Lys-Cys-Cys-Lys-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific DEFS003B
Encompass
SEQUENCE: H-Asp-His-Tyr-Asn-Cys-Val-Ser-Ser-Gly-Gly-Gln-Cys-Leu-Tyr-Ser-Ala-Cys-Pro-Ile-Phe-Thr-Lys-Ile-Gln-Gly-Thr-Cys-Tyr-Arg-Gly-Lys-Ala-Lys-Cys-Cys-Lys-OH (trifluoroacetate salt)(Cys5 and 34 bridge, Cys12 and 27 bridge, Cys17 and 35 bridge)ONE-LETTER SEQUENCE: DHYNCVSSGGQCLYSACPIFTKIQGTCYRGKAKCCK (C5&C34 bridge, C12&C27 bridge, C17&C35 bridge)MOLECULAR FORMULA: C167H256N48O50S6MOLECULAR WEIGHT: 3928.6STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: -Defensin-1, humanRESEARCH AREA: AntimicrobialREFERENCES:K.W. Bensch, M. Raida, H-J. M?gert, P. Schulz-Knappe, and W.-G. Forssmann, FEBS Lett., 368, 331 (1995). (Original Article)R.I. Lehrer and T. Ganz, Ann. N.Y. Acad. Sci., 797, 228 (1996). (Review Article)SKU(s): DEFS-003A, DEFS-003B
Catalog No. 50-210-9189
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.