Learn More
Abcam Beta-Elemene, Anticancer Agent, 204.35 Da, 25 mg
Description
- Molecular weight: 204.35 Da
- Anticancer agent: Induces G2M cell cycle arrest and apoptosis
- Mechanism: Induces caspase-3, -7 and -9 activities; decreases Bcl-2 expression; induces mitochondrial release of cytochrome c; inhibits microtubule polymerization; downregulates uPA, uPAR, MMP-2, and MMP-9 expression
- Effects: Shows antiproliferative and antitumor effects
- Composition: 100% β-Elemene
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Target Molecule | Beta-Elemene |
| Buffer | 100% beta-elemene |
| Content And Storage | Store at 4°C. |
| Formulation | 100% β-Elemene |
| Quantity | 25 mg |
| For Use With (Application) | Anticancer research, Apoptosis study, Cell cycle analysis |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.