Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Beta-Elemene, Anticancer Agent, 500 mg

Catalog No. 502705421
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5421 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5421 Supplier Abcam Supplier No. AB145171500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Anticancer agent that induces G2M cell cycle arrest and apoptosis.

  • Molecular Weight: 204.35 Da
  • Anticancer agent
  • Mechanism of action: Induces G2M cell cycle arrest and apoptosis; induces caspase-3, -7 and -9 activities; decreases Bcl-2 expression; induces mitochondrial release of cytochrome c; inhibits microtubule polymerization; downregulates uPA, uPAR, MMP-2, and MMP-9 expression
  • Biological effects: Shows antiproliferative and antitumor effects
  • Storage: Store at +4°C
  • Composition: 100% β-Elemene

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Beta-Elemene
Shipping Condition Ambient
Form Liquid
Buffer 100% β-Elemene
Content And Storage Store at 4°C.
Formulation 100% β-Elemene
Quantity 500 mg
For Use With (Application) Anticancer research, Antiproliferative studies, Antitumor studies
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.