Learn More
Abcam Beta Galactosidase Assay Kit, LacZ Blue, 500 Tests
Description
- Application: Flow cytometry
- Quantitative capability: Yes
- Cited in: More than 7 publications
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Substrate | LacZ |
| Target Molecule | Beta-Galactosidase |
| No. of Reactions | 500 Tests |
| For Use With (Equipment) | Flow Cytometer |
| Product Type | Western Blot Kit |
| Detection Method | Fluorescence |
| Detection Location | In-Cell Detection |
| Format | Kit |
| Quantity | 500 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.