Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Beta Galactosidase Detection Kit, Fluorometric, Cell-Based, 500 Tests

Catalog No. 502582286
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-2286 500 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-2286 Supplier Abcam Supplier No. AB176721500TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rapid fluorometric assay for the quantitative detection of Beta-galactosidase activity in cell samples within 30 minutes.

  • Assay time: 30 min
  • Detection method: Fluorescent
  • Sensitivity: ≥ 0.3 mU/well
  • Assay type: Cell-based
  • Sample types: Suspension cells, Adherent cells
  • Results type: Quantitative
  • Applications: Enzyme activity assay
  • Cited in: > 6 publications (8 citations)

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Beta-galactosidase
Run Time 30 min.
Substrate Beta-galactosidase
No. of Reactions 500 Tests
Detection Location In-Cell Detection
For Use With (Equipment) Microplate Reader, Fluorescence Microscope
Format Kit
Detection Method Fluorescence
Sample Type Suspension cells, Adherent cells
Quantity 500 Tests
For Use With (Application) Enzyme activity assay
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.