Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Beta-Glucuronidase

Catalog No. MP215228450 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mL
2 mL
10 mL
50 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215228450 50 mL
ICN15228401 1 mL
ICN15228410 10 mL
ICN15228402 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MP215228450 Supplier MP Biomedicals™ Supplier No. 0215228450
Add to Cart
Add to Cart

For analysis of steroid-gluceronides

Beta-Glucuronidase, [9001-45-0]
  • For analysis of steroid-gluceronides
  • In 65% saturated ammonium sulfate
  • Activity: ≤5000 u/mL
  • Unit Definition: Amount that liberates 1.0μg of phenolphthalein from phenolphthalein glucuronide per hour at pH 5.0 and 37°C
TRUSTED_SUSTAINABILITY

Specifications

Concentration beta-Glucuronidase: >50,000 U/mL, Sulfatase: 5,000 U/mL
Color Brown
For Use With (Application) Enzymatic hydrolysis of Glucuronides from Serum Plasma and other fluids prior to analysis by Enzyme immunoassay, mass spectrometry, GC, HPLC
Content And Storage 4°C
Source Helix pomatia
Quantity 50 mL
Product Type beta-Glucuronidase
Inhibitors D-glucuronic Acid, D-galacturonic Acid and D-glucaro-1,4-lactone
Form Solution
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.