Learn More
Abcam Beta-Hydroxypyruvic Acid, >95% Purity, 25 mg
Description
- Molecular weight: 104.06 g/mol
- Purity: >95%
- Biological function: Metabolite involved in the pathway of carbon in photorespiration
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | beta-Hydroxypyruvic Acid |
| Substrate Properties | Chemical Substrate |
| Quantity | 25 mg |
| For Use With (Application) | Photorespiration |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.