Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Beta Lactoglobulin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119523
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119523 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119523 Supplier Invitrogen™ Supplier No. PA5119523
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Beta-lactoglobulin is a major protein found in dairy products and is not present in human milk. It has been identified as a potential milk allergen, and is responsible for transport of both iron and hydrophobic molecules.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Beta Lactoglobulin
Applications ELISA, Functional Assay, Immunoelectrophoresis, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene PAEP
Gene Accession No. P02754
Gene Alias beta lactoglobulin C; beta lactoglobulin D; beta-lactoglobulin; Beta-LG; BLG; bLG C; Bos d 5; lactoglobulin, beta; LGB; PAEP; progestagen-associated endometrial protein
Gene Symbols PAEP
Host Species Rabbit
Immunogen Beta lactoglobulin extract from bovine milk.
Purification Method Antigen affinity chromatography
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 280838
Target Species Bovine
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.