Learn More
Medchemexpress LLC Beta muricholic acid | 2393-59-1 | MFCD00271403 | 99.9% | C24H40O5 | 10MG

Supplier: Medchemexpress LLC HY133707R10MG
β-Muricholic acid (CAS 2393-59-1) is an analytical standard intended for research and analytical applications. Supplied in a small-mass pack, it is used as a calibration and reference standard for bile acid and lipid metabolism studies, and for method development and QC in LC-MS and GC-MS.
- High purity (HPLC) 99.9%
- Molecular formula C24H40O5; MW 408.57 g/mol
- Pack size: 10 mg for preparing stock solutions
- Suitable for LC-MS and GC-MS calibration and quality control
- Relevant for bile acid, cholesterol gallstone, and NAFLD research
- Storage: 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.