Learn More
Abcam Beta-Nicotinamide Adenine Dinucleotide, P2Y11 Agonist, Solid, 1 g
Description
- Molecular Weight: 663.4 Da
- Purity: >95%
- Form: Solid
- Compound Activity: P2Y¹¹ agonist
- Storage Buffer: Constituents: 100% Nadide
- Storage Conditions: Store at -20°C
- Shipping Conditions: Ambient - Can Ship with Ice
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | P2Y11 |
| Shipping Condition | Ambient |
| Form | Solid |
| Buffer | 100% Nadide |
| Content And Storage | Store at -20°C. |
| Formulation | 100% Nadide |
| Quantity | 1 g |
| For Use With (Application) | P2Y11 agonist |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.