Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Beta-Nicotinamide Adenine Dinucleotide, P2Y11 Agonist, Solid, 25 mg

Catalog No. 502584606
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-4606 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-4606 Supplier Abcam Supplier No. AB12040325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Highly pure P2Y11 receptor agonist with >95% purity.

  • P2Y11 agonist
  • Molecular Weight: 663.4 Da
  • Purity: >95%
  • Form: Solid
  • CAS Number: 53-84-9
  • Constituents: 100% Nadide

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Substrate Properties Chemical Substrate
Target Molecule P2Y11
Buffer 100% Nadide
Form Solid
Content And Storage Store at -20°C.
Formulation 100% Nadide
Quantity 25 mg
For Use With (Application) P2Y11 agonist
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.