Learn More
Abcam Beta-Nicotinamide Adenine Dinucleotide, P2Y11 Agonist, Solid, 250 mg
Description
- P2Y11 agonist
- Purity: >95%
- Molecular Weight: 663.4 Da
- Form: Solid
- Constituents: 100% Nadide
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | P2Y11 (Receptor) |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Content And Storage | Store at -20°C. |
| Buffer | 100% Nadide |
| Form | Solid |
| Formulation | 100% Nadide |
| Quantity | 250 mg |
| For Use With (Application) | P2Y11 receptor agonist |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.