Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beta-Nicotinamide Adenine Dinucleotide Reduced Disodium Salt, MP Biomedicals

Catalog No. ICN10116810 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
1 g
5 g
10 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10116810 10 g
ICN16005050 50 mg
ICN16005083 250 mg
ICN16005090 500 mg
ICN16005091 1 g
ICN16005094 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. ICN10116810 Supplier MP Biomedicals, Inc Supplier No. 0210116810
Add to Cart
Add to Cart

Chemical Identifiers

CAS 606-68-8
Molecular Formula C21H27N7Na2O14P2
Molecular Weight (g/mol) 709.41
MDL Number MFCD00036200
InChI Key QRGNQKGQENGQSE-WUEGHLCSSA-L
Synonym beta-nadh disodium salt
PubChem CID 131673989
IUPAC Name disodium [(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl ({[(2R,3S,4R,5R)-5-(3-carbamoyl-1,4-dihydropyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
SMILES [Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O

Specifications

Quantity 10 g
Formula Weight 709.4
Chemical Name or Material Beta-Nicotinamide Adenine Dinucleotide Reduced Disodium Salt
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.