Learn More
Sigma Aldrich Fine Chemicals Biosciences MES hydrate 99.5 titration25KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences M825025KG
Encompass
MES is one of the Good buffers the most extensively used biological buffers. MES is a zwitterionic N-substituted aminosulfonic acid with a morpholinic ring. It does not form complexes with majority of the metals used in environmental and biological studies. It is easily soluble in water and has minimum lipid solubility making it impermeable to membranes.
Catalog No. 50-291-4458
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.