Learn More
Cayman Chemical β-Nicotinamide Mononucleotide, 1094-61-7, 10 mg
SDP
Supplier:  Cayman Chemical 1641110
Encompass_Preferred
Molecular Formula C11H15N2O8P, Purity ≥95%, Formula Weight 334.2, Synonyms: β-NMN; An intermediate in the biosynthesis of NAD+ that has been used to enhance NAD biosynthesis and glucose-stimulated insulin secretion in Nampt+/- mice and high-fat diet-induced type 2 diabetes mice.
Catalog No. NC1019639
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.