Learn More
Market Lab Beta Splash Shield, Small, 11.5"W x 7.5"D x 15"H
Supplier:  Market Lab 14600
Encompass_Preferred
Built-in protection against Beta Isotopes. 0.5" acrylic effectively reduces bremsstrahlung production. Durable acrylic shielding provides effective protection for P-32 and other beta emitters. Designed for easy use and viewing. 11.5" W x 7.5" D x 15" W.
Catalog No. NC1614252