Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Tetralone 95.0+%, TCI America™
SDP

Catalog No. T116725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T116725G 25 g
T11675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T116725G Supplier TCI America Supplier No. T116725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 530-93-8
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.189
MDL Number MFCD00001727
InChI Key KCKZIWSINLBROE-UHFFFAOYSA-N
Synonym 2-tetralone, beta-tetralone, 1,2,3,4-tetrahydro-2-naphthalenone, 3,4-dihydronaphthalen-2 1h-one, 1,2,3,4-tetrahydronaphthalen-2-one, 2 1h-naphthalenone, 3,4-dihydro, .beta.-tetralone, tetralin-2-one, 3,4-dihydro-2 1h-naphthalenone, 3,4-dihydro-2-oxonaphthalene
PubChem CID 68266
IUPAC Name 3,4-dihydro-1H-naphthalen-2-one
SMILES C1CC2=CC=CC=C2CC1=O

Specifications

Color Yellow
Boiling Point 240°C
Quantity 25 g
Formula Weight 146.19
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material beta-Tetralone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.