Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Recommended positive controls: Jurkat, Raji, NCI-H929, U87-MG, SK-N-SH, IMR32, SK-N-AS, C8D30, NIH3T3, BCL-1, Raw 264.7, C2Cl2, Drosophila lysate , rat brain. Predicted reactivity: Mouse (100%), Rat (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Tubulin is the major constituent of microtubules. It binds two moles of GTP, one at an exchangeable site on the beta chain and one at a non-exchangeable site on the alpha-chain.
Specifications
Specifications
| Antigen | beta Tubulin 2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | TUBB2A |
| Gene Accession No. | P61857, P85108, Q13885, Q3KRE8, Q7TMM9, Q9BVA1, Q9CWF2 |
| Gene Alias | 2410129E14Rik; 2t; b2 tubulin; B2t; bA506K6.1; beta 2 tubulin; BETA 85D; beta tubulin; beta(2)Tu; beta(2)Tub; beta(2)Tubulin; beta;Tub85D; beta[[2]]-tubulin; BETA2; beta2 tubulin; beta2t; beta2tub; beta2-tub; beta2tubulin; Beta-2-tubulin; beta2-tubulin; beta85D; betaTub; beta-tub; betaTub2; BetaTub85D; beta-tub85D; betaTub85D-PA; beta-Tubulin; beta-Tubulin 85D; beta-Tubulin at 85D; beta-tubulin T beta15 (aa 1-445); betaTubulin85D; beta-tubulin85D; brdp; CDCBM5; CG9359; CG9359-PA; class II beta-tubulin isotype; class IIa beta-tubulin; class IIb beta-tubulin; D.m.BETA-85D; dJ40E16.7; Dmbeta2; Dmel\CG9359; Dmel_CG9359; DTB4; GH02051p; M(beta)2; ms(3)KK[D]; PMGYSA; RGD1309427; t beta-15; testis-specific beta-tubulin; Tub; Tub2A; TUBB; TUBB2; TUBB2A; tubb2a {ECO:0000250; Tubb2b; TubB85D; tubulin; tubulin beta; tubulin beta 2A class IIa; tubulin beta 2B class IIb; Tubulin beta class IIa; Tubulin beta-2 chain; Tubulin beta-2A chain; tubulin beta-2B chain; tubulin, beta 2; tubulin, beta 2A class IIa; tubulin, beta 2B class IIb; tubulin, beta polypeptide; tubulin, beta polypeptide 2; tubulin, beta polypeptide paralog; UniProtKB:Q7TMM9} |
| Gene Symbols | betaTub85D, TUBB2A, TUBB2B |
| Show More |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
