Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beta tubulin cofactor D Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988067
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8067 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8067 Supplier Bioss Supplier No. BS7849R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cofactor D is one of four proteins (cofactors A, D, E, and C) involved in the pathway leading to correctly folded beta-tubulin from folding intermediates. Cofactors A and D are believed to play a role in capturing and stabilizing beta-tubulin intermediates in a quasi-native confirmation. Cofactor E binds to the cofactor D/beta-tubulin complex; interaction with cofactor C then causes the release of beta-tubulin polypeptides that are committed to the native state.

Specifications

Antigen Beta tubulin cofactor D
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Tbcd
Gene Accession No. Q9BTW9
Gene Alias 2310057L06Rik; A030005L14Rik; Beta-tubulin cofactor D; KIAA0988; mKIAA0988; PP1096; SSD1; SSD-1; TBCD; TFCD; tubulin folding cofactor D; Tubulin-folding cofactor D; Tubulin-specific chaperone D
Gene Symbols Tbcd
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TBCD/Beta tubulin cofactor D.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6904
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.