Learn More
ACROBiosystems Recombinant Protein;Human beta 2-Microglobulin / B2M Protein;HEK293;100UG;B2M,B2M,Beta-2 microglobulin
Supplier:  ACROBiosystems B2MH5225100UG
Encompass_Preferred
Recombinant Protein;Human beta 2-Microglobulin / B2M Protein;100UG;Human B2M, His Tag (B2M-H5225) is expressed from human 293 cells (HEK293). It contains AA Ile 21 - Met 119 (Accession # AAH32589).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 12.6 kDa. The protein migrates as 13 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-9014
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.