Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BetaBasic™ CN Particle Size Guard Cartridges, 5μm Particle Size

Catalog No. p-4523770 Shop All Thermo Scientific Products
Change view
Click to view available options
Diameter (Metric):
2.1 mm
4 mm
Length (Metric):
10 mm
Particle Size:
5 μm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Length (Metric) Particle Size
71705014001 4 mm 10 mm 5 μm
71705012101 2.1 mm 10 mm 5 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 71705014001 Supplier Thermo Scientific™ Supplier No. 71705014001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thermo Scientific™ BetaBasic™ CN Particle Size Guard Cartridges are available in limited dimensions. Please contact Customer Service for more details.

  • The 10mm design offers maximum protection with minimal increase in retention
  • Fit Thermo Scientific UNIGUARD™ direct connection and stand alone holders
  • Highly efficient and reproducible Thermo Scientific columns
  • 150Å pore size for small molecules, peptides and protein digests
  • Superb pH stability
TRUSTED_SUSTAINABILITY

Specifications

Carbon Load 5%
pH 2 to 9
Particle Size 5 μm
Length (Metric) 10 mm
Pore Size 150Å
Surface Area 200 m2/g
Diameter (Metric) 4 mm
For Use With (Equipment) Uniguard Direct-connection and Stand Alone Holder
Endcapped Yes
USP Type L10
Product Line Beta
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.