Learn More
Penn Veterinary Supply Inc Betadine (Povidone-Iodine) Solution | 16 Fl Oz
SDP
Supplier:  Penn Veterinary Supply Inc PVS15516
Encompass
Povidone-iodine | Purity: 10% | 25655-41-8 | 16oz | BETADINE Solution Veterinary is an aqueous solution of 5% povidone-iodine developed as a topical microbicide for use as an anti-infective agent in procedures at animal hospitals and veterinary clinics, including postoperative application to incisions. It is indicated for preparation of skin prior to surgery and for degerming skin, wounds, and mucous membranes in various infection control procedures.
Catalog No. NC0158124
Due to product restrictions, please to purchase or view availability for this product.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.