Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Betamethasone Valerate, Glucocorticoid, 476.6 Da, 100 mg
Description
- Molecular Weight: 476.6 Da
- Purity: >98%
- Form: Solid
- Class: Glucocorticoid; immunomodulator
- Mechanism: Major steroid hormone; exhibits anti-inflammatory and immunosuppressive effects
- Activity: Active in vivo
- Chemical Name: 9-fluoro-11,21-dihydroxy-16-methyl-3,20-dioxopregna-1,4-dien-17-yl valerate
- CAS Number: 2152-44-5
- Storage: Store at ambient temperature
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Target Molecule | Glucocorticoid |
| Content And Storage | Store at ambient temperature. |
| Form | Solid |
| Formulation | 100% 9-fluoro-11,21-dihydroxy-16-methyl-3,20-dioxopregna-1,4-dien-17-yl valerate |
| Quantity | 100 mg |
| For Use With (Application) | Immunomodulation, Inflammation research |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?