Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Betamethasone Valerate, Glucocorticoid, 476.6 Da, Solid, 25 mg

Catalog No. 502706717
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6717 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6717 Supplier Abcam Supplier No. AB14128025MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent glucocorticoid with anti-inflammatory and immunosuppressive effects.

  • MW: 476.6 Da
  • Purity: >98%
  • Form: Solid
  • Glucocorticoid: Major steroid hormone with anti-inflammatory and immunosuppressive effects.
  • Active in vivo.
  • Storage: Store at ambient temperature.
  • Shipping: Can ship with ice.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Glucocorticoid
Substrate Properties Chemical Substrate
Shipping Condition Ambient
Content And Storage Store at ambient temperature.
Form Solid
Quantity 25 mg
For Use With (Application) Immunomodulation, Inflammation research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.