Learn More
Kyfora Bio Beta-sitosterol 5 MG
Supplier: Kyfora Bio 144955
Beta-sitosterol is a naturally occurring sterol lipid that is growing in the bioprocessing space for its ability to improve lipid nanoparticle (LNP) uptake efficiency. LNPs are well known for facilitating the safe and effective delivery of mRNA to target cells, thus enabling therapeutic action. Inclusion of beta-sitosterol in LNP formulations has been shown to increase transfection efficiency rates both in cell cultures and in vivo, leading to particular interest in the use of the lipid in mRNA vaccine development. Kyfora Bio's high purity, synthetic, cell culture grade beta-sitosterol can be used in conjunction with our other lipid products to provide reliable performance for a variety of bioprocessing applications.Key application(s): LNP, Gene delivery, Drug Delivery, Transfection, mRNA vaccine
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.