Learn More
Apexbio Technology LLC Betaxolol 63659-18-7 10mg
SDP
Supplier:  Apexbio Technology LLC B135210
Encompass_Preferred
Betaxolol (CAS 63659-18-7) is a cardioselective 1-adrenergic receptor antagonist that inhibits adrenergic stimulation in cardiac tissue By selectively blocking 1-adrenergic receptors betaxolol modulates cardiovascular responses associated with sympathetic nerve activity In preclinical research betaxolol administered intraperitoneally at 5 mg/kg at defined intervals during cocaine withdrawal has been utilized to investigate its effects on neurobehavioral adaptations following chronic cocaine exposure This compound is widely employed in studies probing -adrenergic involvement in cardiovascular and neuropharmacological processes
Catalog No. 50-000-91385
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.