Learn More
Medchemexpress LLC HY-101519 5mg Medchemexpress, BETd-260 CAS:2093388-62-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1015195MG
Encompass_Preferred
Medchemexpress, HY-101519 5mg BETd-260 CAS:2093388-62-4 BETd-260 is a potent BET degrader based on PROTAC technology, with as low as 30 pM against BRD4 protein in RS4;11 leukemia cell line. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2156
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.