Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tingley™ Better Grade PVC Work Boots

Catalog No. 19161290 Shop All Tingley Rubber Co. Products
Change view
Click to view available options
Size:
5
7
9
10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-161-290 5
19-022952 9
19-022951 7
19-022946 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-161-290 Supplier Tingley™ Supplier No. 5124405

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Blend of high-quality PVC resins, low-temperature plasticizers, and nitrile rubber creates work boots that remain supple even in cold weather

Withstand abrasion, and provide resistance to chemicals such as alcohols, organic and inorganic acids, salts, and alkalis. 100% waterproof. Ideal for agriculture, industrial, and food processing applications.

  • 15 in. high (38cm)
  • Brown upper with crepe sole
  • Cleated outsole spits out debris for excellent gripping action
  • Injection molded construction for 100% waterproof protection
  • Flexible upper material stays supple in cold temperatures to make walking easier
  • Outsole material is soft for excellent slip resistance and absorbs shock when walking
  • Removable polypropylene insole wicks perspiration to help keep feet dry and comfortable

Compliance

Steel-toe work boots meet ANSI Z41 PT99 M/I/75/C/75.

Specifications

Certifications/Compliance ANSI Z41 PT99 M/I/75/C/75.
Color Brown
For Use With (Application) Ideal for agriculture, industrial, and food processing applications.
Height (English) 15 in.
Height (Metric) 38 cm
Material PVC
Size 5
Toe Material Steel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.