Learn More
Cayman Chemical MarbofloxacInd8hydrochLRI 5mg
SDP
Supplier:  Cayman Chemical 332735 MG
Encompass_Preferred
A Kir2 channel inhibitor (IC50s 1.8 2.8 29 and 2.6 UM for Kir2.1 Kir2.2 Kir2.3 and Kir2.6 respectively) selective for Kir2 channels over Kir1.1 and Kir4.1 channels (IC50s 33 and 76 UM respectively) prevents the development of dynamic but not punctate mechanical allodynia in a mouse model of spared nerve injury at 10 and 30 nmol/animal
Catalog No. 50-294-8442
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.