Learn More
Med Vet International Beveled Single Frosted Slides, One End, One Side
Supplier:  Med Vet International 8205
Encompass_Preferred
These beveled edge slides are made of water white glass and have clipped corners (1mm). 75x25mm (3?x1?) approx. 1mm thick. 72pcs per box
Catalog No. 50-304-3142
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.